Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFC4 Monoclonal Antibody (OTI1A8)
GREENER_CHOICE

Catalog No. PIMA526260
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526260 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526260 Supplier Invitrogen™ Supplier No. MA526260
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RFC4 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Human Replication Factor C (RFC) is a hetero-pentameric protein consisting of the p140, p40, p38, p37 and p36 subunits. RFC functions in the loading of the DNA polymerase delta processivity factor (PCNA, Proliferating Cell nuclear Antigen) onto a primed DNA template during DNA synthesis and repair. RFC may also participate in the DNA damage checkpoint response with a PCNA-like hetero-trimer consisting of Hus 1, Rad 1 and Rad 9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFC4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RFC4
Gene Accession No. P35249, Q99J62
Gene Alias A1; A1 37 kDa subunit; Activator 1 37 kDa subunit; activator 1 subunit 4; AI894123; AU040575; EGK_12194; replication factor C (activator 1) 4; replication factor C (activator 1) 4, 37kDa; replication factor C 37 kDa subunit; Replication factor C subunit 4; RFC 37 kDa subunit; RF-C 37 kDa subunit; RFC p37; RFC37; RFC4
Gene Symbols RFC4
Host Species Mouse
Immunogen Full length human recombinant protein of RFC4 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106344, 288003, 478667, 5984
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.