Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566318
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566318 Supplier Invitrogen™ Supplier No. PA566318
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSLCFIHRYYR RSPHRPLAGL HLSP Highest antigen sequence identity to the following orthologs - mouse 80%, rat 76%.

May be involved in N-linked oligosaccharide assembly. May participate in the translocation of oligosaccharide from the cytoplasmic side to the lumenal side of the endoplasmic reticulum membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFT1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFT1
Gene Accession No. Q96AA3
Gene Alias CDG1N; congenital disorder of glycosylation 1N; protein RFT1 homolog; putative endoplasmic reticulum multispan transmembrane protein; RFT1; RFT1 homolog; RFT1, requiring fifty three 1 homolog
Gene Symbols RFT1
Host Species Rabbit
Immunogen Recombinant Human RFT1. Recombinant protein control fragment (Product #RP-106999).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91869
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.