Learn More
Enzo Life Sciences RG-13022 (5mg). CAS: 136831-48-6
SDP
Supplier:  Enzo Life Sciences BMLEI2520005
Encompass_Preferred
RG-13022 is a non-phenolic tyrphostin analog, which inhibits the EGF receptor (IC50=5 µM) and PDGF receptor tyrosine kinases, and is long-acting. It inhibits EGF-stimulated HER14 cell proliferation (IC50=1 µM) as well as tumor growth in vivo. It inhibits both EGF- and PDGF-stimulated DNA synthesis in human glioma cell lines. RG-13022 is non-phenolic and therefore possesses less antioxidant activity than other tyrphostins. Alternative name: 3,4-Dimethoxy-alpha-(3-pyridyl)-(Z)-cinnamonitrile. Purity: ≥98% (TLC). Solubility: Soluble at 25mg/ml in DMSO or 100% ethanol. Long Term Storage: Ambient.
Catalog No. 50-201-0637
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.