Learn More
Abcam RGD Peptide, Arg-Gly-Asp, Cell Adhesion Sequence, Solid, 10 mg
Description
- Molecular weight: 346.34 Da
- Purity: >95%
- Form: Solid
- Composition: 100% RGD
- Citation count: 8
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | RGD |
| Substrate Properties | Peptide-based Substrate |
| Form | Solid |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Quantity | 10 mg |
| For Use With (Application) | Cell adhesion |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.