Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam RGD Peptide, Arg-Gly-Asp Sequence, >95% Purity, Solid, 25 mg

Catalog No. 502706205
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6205 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6205 Supplier Abcam Supplier No. AB14269825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly pure cell adhesion peptide with >95% purity.

  • Molecular Weight: 346.34 Da
  • Purity: >95%
  • Form: Solid
  • Constituents: 100% RGD
  • Citation Count: 8

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Form Solid
Content And Storage Store at -20°C.
Shipping Condition Ambient
Substrate Properties Peptide-based Substrate
Target Molecule RGD (Arg-Gly-Asp)
Quantity 25 mg
For Use With (Application) Cell adhesion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.