Learn More
Cayman Chemical RGFP966, 1357389-11-7, MFCD26960954, 1mg
SDP
Supplier:  Cayman Chemical 169171
Encompass_Preferred
Molecular Formula: C21H19FN4O, Purity: ≥98%, Formula Weight: 362.4, Formal Name: (2E)-N-(2-amino-4-fluorophenyl)-3-[1-(3-phenyl-2-propen-1-yl)-1H-pyrazol-4-yl]-2-propenamide; HDAC3 is highly expressed in brain, and its activity has been associated with negative regulation of learning and memory processes.
Catalog No. NC1612468
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.