Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS1 Recombinant Rabbit Monoclonal Antibody (HL1402)
GREENER_CHOICE

Catalog No. PIMA547033 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547033 Supplier Invitrogen™ Supplier No. MA547033
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

This gene encodes a member of the regulator of G-protein signaling family. This protein is located on the cytosolic side of the plasma membrane and contains a conserved, 120 amino acid motif called the RGS domain. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1402
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RGS1
Gene Accession No. Q08116, Q9JL25
Gene Alias 1R20; B-cell activation protein BL34; BL34; Early response protein 1R20; epididymis secretory protein Li 87; HEL-S-87; IER1; immediate-early response 1, B-cell specific; IR20; Regulator of G-protein signaling 1; regulator of G-protein signalling 1; RGS1
Gene Symbols RGS1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RGS1. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50778, 5996
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.