Learn More
Invitrogen™ RGS11 Polyclonal Antibody

Description
Suggested positive control: human and rat brain tissue or protein. Immunogen sequence: SADFHKREIE YFRKALG.
Specifications
Specifications
| Antigen | RGS11 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | RGS11 |
| Gene Accession No. | O94810, P49807, Q9Z2H1 |
| Gene Alias | AC004754.2; C78048; regulator of G-protein signaling 11; regulator of G-protein signalling 11; RGS11; RS11 |
| Gene Symbols | RGS11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.