Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566919 Supplier Thermo Scientific Supplier No. PA566919
Only null left
Add to Cart
Add to Cart

RGS12 Polyclonal Antibody

RGS12 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS12
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RGS12
Gene Accession No. O14924
Gene Alias 1200016K18Rik; 4632412M04Rik; AI481290; DKFZp761K1617; DKFZp761K1817; E130309H11; regulator of G-protein signaling 12; regulator of G-protein signalling 12; Rgs12; RP11-529E10.1
Gene Symbols RGS12
Host Species Rabbit
Immunogen Recombinant Human RGS12. Recombinant protein control fragment (Product #RP-107572).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6002
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.