Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS16 Monoclonal Antibody (OTI4F9), TrueMAB™, OriGene
SDP

Catalog No. 502540545
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0545 30 μL
50-167-4807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0545 Supplier OriGene Supplier No. TA504033S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RGS16 Monoclonal antibody specifically detects RGS16 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen RGS16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4F9
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RGS16
Gene Accession No. O15492
Gene Alias A28-RGS14; A28-RGS14P; hRGS-r; regulator of G-protein signaling 16; regulator of G-protein signalling 16; Retinally abundant regulator of G-protein signaling; retinal-specific RGS; Rgs14; RGS16; RGSR; RGS-R
Host Species Mouse
Immunogen Full length human recombinant protein of human RGS16 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6004
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.