Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675754
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-754 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-754 Supplier Invitrogen™ Supplier No. PA141189
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: human heart lysate. Immunogen sequence homology: mouse (86%) and rat (86%).

Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 2 belongs to this family. The protein acts as a mediator of myeloid differentiation and may play a role in leukemogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene RGS2
Gene Accession No. O08849, P41220, Q9JHX0
Gene Alias cell growth-inhibiting gene 31 protein; cell growth-inhibiting protein 31; G0 to G1 switch regulatory 8, 24kD; G0/G1 switch regulatory protein 8; G0S8; GIG31; GOS8; regulator of G protein signaling 2; regulator of G protein signaling 2; regulator of G-protein signaling 2; regulator of G-protein signaling 2; regulator of G-protein signaling 2, 24kDa; regulator of G-protein signaling protein 2; regulator of G-protein signalling 2, 24kDa; Rgs2
Gene Symbols RGS2
Host Species Rabbit
Immunogen A synthetic peptide within amino acid residues 1-50 of human RGS2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19735, 5997, 84583
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.