Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA528162
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528162 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528162 Supplier Thermo Scientific Supplier No. PA528162

Rabbit Polyclonal Antibody

PA5-28162 targets RGS2 in IF and WB applications and shows reactivity with Human samples. The PA5-28162 immunogen is recombit fragment corresponding to a region within amino acids 1 and 211 of Human RGS2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 2 belongs to this family. The protein acts as a mediator of myeloid differentiation and may play a role in leukemogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS2
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene RGS2
Gene Accession No. P41220, O08849
Gene Alias G0S8, G0 to G1 switch regulatory 8, 24kD, G0/G1 switch regulatory protein 8, cell growth-inhibiting gene 31 protein, cell growth-inhibiting protein 31, regulator of G-protein signaling 2
Gene Symbols RGS2
Host Species Rabbit
Immunogen Recombit fragment corresponding to a region within amino acids 1 and 211 of Human RGS2
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19735, 5997
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.