Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584835 Supplier Invitrogen™ Supplier No. PA584835
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPQLSQDNQE CLQKHFSRPS IWTQFLPLFR AQRYNTDIHQ ITENEGDLRA VP Highest antigen sequence indentity to the following orthologs: Mouse - 47%, Rat - 30%.

Regulator of G protein signaling (RGS) proteins are regulatory and structural components of G protein-coupled receptor complexes. RGS proteins are GTPase-activating proteins for Gi (see GNAI1; MIM 139310) and Gq (see GNAQ; MIM 600998) class G-alpha proteins. They accelerate transit through the cycle of GTP binding and hydrolysis and thereby accelerate signaling kinetics and termination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS20
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RGS20
Gene Accession No. O76081
Gene Alias 2900073E09Rik; G(z)GAP; gz-GAP; gz-selective GTPase-activating protein; Regulator of G-protein signaling 20; regulator of G-protein signaling 20 variant 2; regulator of G-protein signaling Z1; regulator of G-protein signalling 20; regulator of Gz-selective protein signaling 1; Rgs20; Rgsz1; ZGAP1
Gene Symbols RGS20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RGS20. Recombinant protein control fragment (Product #RP-108248).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8601
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.