Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ RGS5 Protein
SDP

Catalog No. NB813458 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB813458 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB813458 Supplier R&D Systems™ Supplier No. NBP1724140.5MG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-181 of Human RGS5 The Recombinant Human RGS5 Protein is derived from E. coli. The Recombinant Human RGS5 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 0.1M NaCl, 1mM DTT
Molecular Weight (g/mol) 23.5kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Human
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name RGS5
Conjugate Unconjugated
Product Line Novus
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.