Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675536
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-536 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-536 Supplier Invitrogen™ Supplier No. PA141407
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: human and mouse brain tissue or protein.

Members of the RGS (regulator of G protein signaling) family, such as RGS6, modulate G protein function by activating the intrinsic GTPase activity of the alpha (guanine nucleotide-binding) subunits (Seki et al., 1999 [PubMed 10083744]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene RGS6
Gene Accession No. P49758, P49801, Q9Z2H2
Gene Alias DKFZp313G1241; FLJ43552; GAP; H_DJ1108A12.1; MGC142132; regulator of G-protein signaling 6; regulator of G-protein signalling 6; RGS6; S914; WUGSC:H_DJ0283M22.1; WUGSC:H_DJ1108A12.1
Gene Symbols RGS6
Host Species Rabbit
Immunogen Synthetic peptides corresponding to amino acids 182-198 and 410-426 of human RGS6.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50779, 54295, 9628
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.