Learn More
Biotium RH414
Supplier:  Biotium 61016
Encompass_Preferred
The styryl dye RH414 is a fast-responding potentiometric probe that is primarily used for functional imaging of neurons. Excitation/emission data is for the dye in methanol. In cell membranes, the spectra of styryl dyes are typically blue-shifted by as much as 20 nm for absorption or excitation and 80 nm for emission. Properties: Ex/Em (MeOH) = 532/716 nm; Orange red soluble in H2O or DMSO; Store at -20°C and protect from light; C28H43Br2N3; MW: 581.48; [161433-30-3]
Catalog No. 50-196-4737
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.