Learn More
BD Rhapsody™ Mouse TCR/BCR Next Amplification Kit
Catalog No. BDB940513 Shop All BD Biosciences Products
Catalog No. BDB940513 Supplier BD Supplier No. 667059
Add to Cart
Add to Cart
The next generation of BD Rhapsody™ TCR/BCR Multiomic Assays reveal immune repertoire richness with great sensitivity alongside simultaneous single-cell gene and protein expression analyses.
Validated on our new Enhanced Cell Capture Beads V3, BD Rhapsody™ TCR/BCR Next Multiomic Assays enable a highly sensitive analysis of full-length TCR/BCR repertoires with robust workflows for human and mouse. Key features include:
  • Superior full-length TCR and BCR pairing metrics
  • Multiomics-enabled for simultaneous immune profiling alongside protein and gene expression analyses in the same cell
  • Compatibility with preserved cells and BD™ OMICS-Guard Sample Preservation Buffer
  • Intuitive data analysis pipeline that provides unbiased insights about the biology of your samples
Kit provides the necessary reagents to prepare single-cell full-length Mouse TCR and BCR libraries that are compatible with both BD Rhapsody™ Whole Transcriptome Analysis (WTA) and BD Rhapsody™ Targeted mRNA assays.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mouse TCR/BCR Next Amplification Kit