Learn More
Description
Validated on our new Enhanced Cell Capture Beads V3, BD Rhapsody™ TCR/BCR Next Multiomic Assays enable a highly sensitive analysis of full-length TCR/BCR repertoires with robust workflows for human and mouse. Key features include:
- Superior full-length TCR and BCR pairing metrics
- Multiomics-enabled for simultaneous immune profiling alongside protein and gene expression analyses in the same cell
- Compatibility with preserved cells and BD™ OMICS-Guard Sample Preservation Buffer
- Intuitive data analysis pipeline that provides unbiased insights about the biology of your samples
Specifications
Specifications
| Species | Human |
| No. of Samples | 4 Samples |
| Target | mRNA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.