Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHBDF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548602 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548602 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548602 Supplier Invitrogen™ Supplier No. PA548602
  • $588.00
    $485.65 / Each of 1

    Save $102.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

RHBDF2 Polyclonal Antibody for Western Blot, IHC (P)

Regulates ADAM17 protease, a sheddase of the epidermal growth factor (EGF) receptor ligands and TNF, thereby plays a role in sleep, cell survival, proliferation, migration and inflammation. Does not exhibit any protease activity on its own. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBDF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RHBDF2
Gene Accession No. Q6PJF5, Q80WQ6
Gene Alias Inactive rhomboid protein 2; IRHOM2; RHBDF2; RHBDL5; RHBDL6; rhomboid 5 homolog 2; Rhomboid family member 2; rhomboid veinlet-like protein 5; Rhomboid veinlet-like protein 6; TEC; TOC; TOCG
Gene Symbols RHBDF2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 80-109 amino acids from the N-terminal region of human RHBDF2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217344, 79651
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.