Learn More
MilliporeSigma™ Supelco™ RheBuild™ Kit
Catalog No. 111004068 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With:
8125/8126 Valves
Catalog No. For Use With
11-100-4068 8125/8126 Valves
1 options
Catalog No. 11-100-4068 Supplier MilliporeSigma™ Supelco™ Supplier No. 55050U
Add to Cart
Add to Cart
Includes all of the tools and parts you need to repair your Rheodyne injector
The parts included are those most likely to be damaged, worn, or lost. Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.
TRUSTED_SUSTAINABILITY
Specifications
For Use With 8125/8126 Valves
RheBuild is a registered trademark of IDEX Health & Science, LLC