Learn More
MilliporeSigma™ Supelco™ RheFlex™ PEEK Fittings Ferrule Pack, 1/16 in. Tubing
Catalog No. 111011937 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
5/Pk.
Catalog No. Packaging Quantity
11-101-1937 5/Pk.
1 options
Catalog No. 11-101-1937 Supplier MilliporeSigma™ Supelco™ Supplier No. 57692
Add to Cart
Add to Cart
PEEK (polyetheretherketone) fingertight fittings (ferrule pack) are convenient, inert, and biocompatible.
Use these fittings with 1/16 in. O.D. PEEK, stainless steel, titanium, Tefzel, or PTFE tubing. PEEK fittings are compatible with all HPLC solvents (avoid concentrated sulfuric and nitric acids), and can be used at temperatures up to 150 C. Unlike stainless steel ferrules, PEEK ferrules do not permanently lock into place on the tubing. This allows the interchanging of fittings with tubing and columns, from manufacturer to manufacturer (e.g. from Agilent to Waters), and still form a zero dead volume, leak-free connection.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 5/Pk.
Material PEEK
RheFlex is a registered trademark of IDEX Health & Science, LLC