Learn More
MilliporeSigma™ Supelco™ Rheodyne™ Fittings long Nut for 1/16 in. Tubing
Catalog No. 111004885 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
Catalog No. Packaging Quantity
11-100-4885 10/Pk.
1 options
Catalog No. 11-100-4885 Supplier MilliporeSigma™ Supelco™ Supplier No. 58257
Add to Cart
Add to Cart
The long Rheodyne™ nut has a longer hex portion for easier wrench access.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 10/Pk.
Material Stainless Steel
Rheodyne is a registered trademark of IDEX Health & Science, LLC