Learn More
MilliporeSigma™ Supelco™ Rheodyne™ Port Adapter
Catalog No. 111005019 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Rheodyne™ Port Adapter
Catalog No.
11-100-5019 Rheodyne™ Port Adapter
1 options
Catalog No. 11-100-5019 Supplier MilliporeSigma™ Supelco™ Supplier No. 57472
Add to Cart
Add to Cart
Use the Port Adapter for connecting 1/16 in. tubing to a 1/8 in. valve port.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Chromatography Applications
Rheodyne is a registered trademark of IDEX Health & Science, LLC