Learn More
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Catalog No. 03050413
Change view
Click to view available options
For Use With (Equipment):
Rheodyne Injector 7030
Rheodyne Injector 7410
Rheodyne Injector 7413
Rheodyne Injector 8125
Rheodyne Injector 9010
Rheodyne Injector 9125
Rheodyne Injectors 3725/3725i/3725-038/3725i-038
Rheodyne Injectors 7000/7010/7040
Rheodyne Injectors 7000/7010/7040/7067
Rheodyne Injectors 7060/7066
Rheodyne Injectors 7060/7066/9060
Rheodyne Injectors 7125/7126
Rheodyne Injectors 8125/8126
Material:
ETFE
PEEK
Vespel
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material
03-050-413 Rheodyne Injectors 3725/3725i/3725-038/3725i-038 PEEK
03-052-241 Rheodyne Injectors 7000/7010/7040/7067 Vespel
03-052-261 Rheodyne Injectors 7125/7126 Vespel
03-052-244 Rheodyne Injectors 7000/7010/7040 ETFE
03-052-256 Rheodyne Injector 7030 Vespel
03-052-257 Rheodyne Injector 7030 ETFE
03-052-259 Rheodyne Injectors 7060/7066 Vespel
03-052-260 Rheodyne Injectors 7060/7066/9060 ETFE
03-052-264 Rheodyne Injectors 7125/7126 ETFE
03-052-269 Rheodyne Injector 7410 Vespel
03-052-271 Rheodyne Injector 7410 ETFE
03-052-273 Rheodyne Injector 7413 Vespel
03-052-365 Rheodyne Injectors 8125/8126 Vespel
03-052-369 Rheodyne Injector 8125 ETFE
03-052-394 Rheodyne Injector 9010 ETFE
03-052-389 Rheodyne Injector 9125 ETFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 03-050-413 Supplier Thermo Scientific™ Supplier No. 3725018
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Maintain performance of your Thermo Scientific™ Rheodyne™ Sample Injectors for HPLC with these replacement rotor seals.
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Rotor Seals
Material PEEK
Description 3725-018
For Use With (Equipment) Rheodyne Injectors 3725/3725i/3725-038/3725i-038
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.