Learn More
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Description
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.
Specifications
Specifications
| Type | Replacement Rotor Seals |
| Material | Vespel |
| Description | 7410-038 |
| For Use With (Equipment) | Rheodyne Injector 7410 |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.