Learn More
Bon Opus Biosciences Recombinant Rhesus Macaque CD47 (N-6His-Flag) -50ug
SDP
Supplier:  Bon Opus Biosciences C01R50UG
Encompass_Preferred
Recombinant Rhesus macaque Leukocyte Surface Antigen CD47 is produced by our Mammalian expression system and the target gene encoding Gln19-Glu141 is expressed with a 6His, Flag tag at the N-terminus. Expression system: Human Cells. Accession# F7A802. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# C01R
Catalog No. 50-208-0335
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.