Learn More
Specifications
Specifications
| Gene | EDA2R/TNFRSF27/XEDAR |
| Accession Number | NP_001180396.1 |
| Gene ID (Entrez) | 60401 |
| Gene Alias | ectodysplasin A2 receptor, EDA-A2 receptor, EDAA2R, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, TNFRSF27, XEDAR, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Rhesus Macaque |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
