Learn More
Sino Biological Rhesus IL18RAP PAb 100UL 90122-T16-100
Supplier:  Sino Biological 90122T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus IL18RAP (Catalog#90122-C08H; XP_001108108.1; Met1-Arg356). IL18RAP specific IgG was purified by Rhesus IL18RAP affinity chromatography.
Catalog No. 50-240-9180
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.