Learn More
Sino Biological Recombinant Rhesus IL-2R beta/CD122 Protein (ECD, His & AVI Tag), Avi-tag Biotinylated, HPLC-verified
Supplier:  Sino Biological 90328K49HB100
Encompass_Preferred
A DNA sequence encoding the Rhesus IL2RB (NP_001244989.1) (Met1-Asp239) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-3150