Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhesus IL-18 Recombinant Protein

Catalog No. ENRRMIL18I
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENRRMIL18I 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENRRMIL18I Supplier Invitrogen™ Supplier No. RRMIL18I

Recombinant Protein

Mature rhesus macaque IL-18 was expressed in Pichia pastoris, X-33, GenBank Accession # AAK13416, amino acids 37 - 193. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: YFGKLESKLS. Purity is >95% by SDS-PAGE analyses. Activity: The biological activity of recombinant rhesus macaque IL-18 was determined by its ability to induce IFNg production from KG-1 cells in the presence of recombinant mouse IL-12. 270 pg/mL and 470 pg/mL of IFNg are induced by 20 ng/mL and 40 ng/mL of recombinant rhesus macaque IL-18, respectively. Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 0.5 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Control, ELISA, Immunoprecipitation, Western Blot
Formulation PBS with no preservative
Gene ID (Entrez) 574151
Molecular Weight (g/mol) 18.3 kDa
Name Rhesus IL-18
Quantity 20 μg
Source Yeast
Regulatory Status RUO
Gene Alias cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1 g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320
Common Name IL-18
Gene Symbol IL18
Biological Activity ED50 = 20 - 40 ng/mL; determined by the dose-dependent induction of IFN-gamma production from KG-1 cells in the presence or recombinant mouse IL-12.
Conjugate Unconjugated
Species Yeast
Recombinant Recombinant
Sequence Rhesus macaque IL-18, amino acids 37-193 (Accession # AAK13416)
Content And Storage 4°C
Expression System P. pastoris
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.