Learn More
ACROBiosystems Rhesus macaque CA125 (13647-14291), His Tag (CA5-R52H6) is expressed from human 293 cells (HEK293). It contains AA Pro 13647 - Pro 14291 (Accession # XP_015296314.1).
Supplier:  ACROBiosystems CA5R52H6100UG
Encompass_Preferred
The CA125, also known as the MUC16, is a mucin protein that may be found in type I transmembrane or secreted forms that are used monitor the progress of epithelial ovarian cancer therapy. The CA 125 molecule is almost certainly a glycoprotein with a predominance of O-linkages. It is heterogeneous with regard to both size and charge, most likely due to continuous deglycosylation of side chains during its life-span in bodily fluids. It exists as a very large complex (perhaps as much as 4 million daltons) under natural conditions.
Catalog No. 50-223-1517
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.