Learn More
Bon Opus Biosciences Recombinant Rhesus Macaque IL-33 (N-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CR5650UG
Encompass_Preferred
Recombinant Rhesus macaque Interleukin 33 is produced by our E.coli expression system and the target gene encoding Ser112-Ile270 is expressed with a 6His tag at the N-terminus. Expression system: E.coli. Accession# F7DLG4. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CR56
Catalog No. 50-208-0389
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.