Learn More
ACROBiosystems Rhesus macaque TROP-2 1mg
Supplier:  ACROBiosystems TR2R52551MG
Encompass_Preferred
Rhesus macaque TROP-2 Protein Fc Tag (TR2-R5255) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 274 (Accession # XP_001114599.1).
Catalog No. 50-253-4013
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.