Learn More
Cayman Chemical HexacosnoIc ACD methyl Ste 1mg
SDP
Supplier:  Cayman Chemical 280851 MG
Encompass_Preferred
A metabolite of a-tocotrienol that has antioxidant and ferroptosis inhibitory activity inhibits BSO-induced decreases in cell in primary fibroblasts isolated from patients with Friedreichs ataxia or Leigh syndrome (EC50s 24 and 30 nM respectively) inhibits RSL3-induced cell death and lipid peroxidation in primary fibroblasts isolated from patients with PCH6 (EC50s 17 3-21 8 and 33 1-57 6 nM respectively)
Catalog No. 50-294-1703
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.