Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Rhizopus Asexual Reproduction, Whole Mount
SCI_ED

Catalog No. S30280 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30280 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30280 Supplier Eisco™ Supplier No. BS18126
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prepared microscope slide of Rhizopus asexual reproduction. Whole Mount. Rhizopus is a parasitic fungus that can reproduce via sexual or asexual spores. Asexual spores, sporangiospores, develop in sporangium. This slide is an excellent tool to visualize asexual reproduction structures more closely.

  • Pre-cleaned glass slide with ground edges
  • Size: 25 x 77 mm (1 x 3 in.)
  • Professionally stained samples for best visualization
  • Ideal for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows asexual reproduction structures

Specifications

Biosafety Level 1
Concentration 10%
Classification Fungi
Material Glass
Mounting Type Whole Mount
Quantity 1 Ea.
Specimen Type Rhizopus Asexual Reproduction
Size 25 x 77 mm (1 x 3 in.)
Slide Type Prepared Slide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.