Learn More
Santa Cruz Biotechnology RHO A MSE MONO.IGG1 ANTI
Supplier:  Santa Cruz Biotechnology SC418
Encompass
Rho A (26C4), mouse monoclonal IgG1; 200microgram/ml, epitope corresponding to amino acids 120-150 of Rho A of human origin, specific for Rho A of mouse, rat and human origin, Western blotting, immunoprecipitation and immunohistochemistry
Catalog No. NC9780116
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.