Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoA/RhoC Monoclonal Antibody (1B3-4A10)
GREENER_CHOICE

Catalog No. PIMA1123
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1123 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1123 Supplier Invitrogen™ Supplier No. MA1123
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-123 detects GTPases RhoA and RhoC, but does not react with RhoB. MA1-123 has been successfully used in Western blot, immunofluorescence, IHC (P) and immunoprecipitation applications with human and mouse samples.

RhoA is described as a small GTPase protein that regulates the actin cytoskeleton in the formation of stress fibers. RhoA is also included in the related proteins of the Ras superfamily, which involves proteins in the regulation and timing of cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoA/RhoC
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1B3-4A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene RHOC
Gene Accession No. P08134, P61586, P61589, Q62159, Q9QUI0
Gene Alias AI324259; Aplysia ras-related homolog 12; aplysia ras-related homolog 9 (RhoC); aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; ARH12; ARH9; ARHA; Arha1; Arha2; Arhc; h12; H9; oncogene RHO H12; oncogene RHO H9; plysia ras-related homolog A2; Ras family member A; ras homolog 9 (RhoC); ras homolog A1; ras homolog A2; ras homolog family member A; ras homolog family member C; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; ras homolog gene family, member C; RAS-related homolog 9; Rho; Rho cDNA clone 12; Rho cDNA clone 9; Rho family GTPase; RHO12; Rhoa; Rhoc; rhoC GTPase; RHOH12; RHOH9; rho-related GTP-binding protein RhoC; SIG-61; silica-induced gene 61 protein; small GTP binding protein RhoA; small GTP binding protein RhoC; transforming protein RhoA
Gene Symbols RHOA, RHOC
Host Species Mouse
Immunogen Full-length human recombinant RhoA protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117273, 11848, 11853, 295342, 387, 389
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.