Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595631
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595631 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595631 Supplier Invitrogen™ Supplier No. PA595631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, rat brain tissue, rat C6 whole cell, mouse brain tissue, monkey COS-7 whole cell. IHC: human renal cancer tissue, human mammary cancer tissue. ICC/IF: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Rho-related GTP-binding protein (RhoB) mediates apoptosis in neoplastically transformed cells after DNA damage. While not essential for development, it does affect cell adhesion and growth factor signaling in transformed cells. RhoB targets PKN1 to endosomes and is involved in trafficking of the EGF receptor from late endosomes to lysosomes. Also required for stability and nuclear trafficking of AKT1/AKT which promotes endothelial cell survival during vascular development. Serves as a tumor supressor - deletion of RhoB causes tumor formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene RHOB
Gene Accession No. P62745, P62746, P62747
Gene Alias AA017882; Aplysia RAS-related homolog 6; aplysia ras-related homolog B; Arh6; Arhb; h6; MST081; MSTP081; oncogene RHO H6; ras homolog B; ras homolog family member B; ras homolog gene family, member AB; ras homolog gene family, member B; Rho; rho cDNA clone 6; rhoB; RHOH6; Rho-related GTP-binding protein RhoB
Gene Symbols RHOB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of human RHOB (NKKDLRSDEHVRTELARMKQEPVRTDDGRAMAVRIQAYDYLE).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11852, 388, 64373
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.