Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoBTB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA130203
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA130203 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA130203 Supplier Invitrogen™ Supplier No. PA130203
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-30203 detects DBC2 from rat, mouse, human samples.

RHOBTB2 is a member of the evolutionarily conserved RHOBTB subfamily of Rho GTPases. For background information on RHOBTBs, see RHOBTB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoBTB2
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.1% sodium azide
Gene RHOBTB2
Gene Accession No. Q91V93, Q9BYZ6
Gene Alias Dbc2; Deleted in breast cancer 2 gene protein; deleted in breast cancer 2 gene protein homolog; E130206H14Rik; KIAA0717; KIAA0740; mKIAA0717; p83; Rho related BTB domain containing 2; Rho related BTB domain containing protein 2; RHOBTB1; RHOBTB2; Rho-related BTB domain containing 2; rho-related BTB domain-containing protein 2
Gene Symbols RHOBTB2
Host Species Rabbit
Immunogen Peptide mapping to the amino terminal domain of human DBC2.
Purification Method Antigen Affinity Chromatography
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23221, 246710, 306004
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.