Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoC Monoclonal Antibody (109)
GREENER_CHOICE

Catalog No. 01670404
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-404 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-404 Supplier Invitrogen™ Supplier No. MA17670
Only null left
Add to Cart
Add to Cart

Human Monoclonal Antibody

MA1-7670 is a RhoC human Rhesus system antigen IgM antibody MA1-7670 has been successfully used in agglutination applications. The MA1-7670 immunogen is RhoC human Rhesus system protein.

RhoC is part of blood-group antigens that are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoC
Applications Agglutination
Classification Monoclonal
Clone 109
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1.25mM EDTA, 0.3M NaCl and 0.1% sodium azide; pH 7.2
Gene Rhoc
Gene Accession No. P08134
Gene Alias AI324259; aplysia ras-related homolog 9 (RhoC); AR ARHC; Arh9; ARHC; h9; MGC1448; MGC61427; oncogene RHO H9; ras homolog 9 (RhoC); ras homolog family member C; ras homolog gene family, member C; RAS-related homolog 9; Rho; rho cDNA clone 9; RHO MGC1448; RHOC; rhoC GTPase; RHOH9; Rho-related GTP-binding protein RhoC; SIG-61; silica-induced gene 61 protein; small GTP binding protein RhoC
Gene Symbols Rhoc
Host Species Human
Immunogen RhoC human Rhesus system protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389
Target Species Human
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.