Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Rhod-2 AM, Fluorescent Ca2+ Indicator, Solid, 1 mg

Catalog No. 502583314
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3314 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3314 Supplier Abcam Supplier No. AB1427801MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable fluorescent Ca2+ indicator with over 20 citations.

  • Cell permeability: Cell-permeable
  • Binding properties: Binds to Ca²± (Kd = 570 nM)
  • Optical properties: Fluoresces yellow/orange upon Ca²± binding; Excitation/Emission: 552 nm/581 nm
  • Purity: > 93%
  • Solubility: Soluble in DMSO to 1 mg/mL
  • Citations: Cited in over 20 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Calcium
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Label or Dye Fluorescent Probe
Form Solid
Content And Storage Store at -20°C.
Detection Location In-Cell Detection
Detection Method Fluorescence
Quantity 1 mg
For Use With (Application) Ca2+ detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.