Learn More
Description
MA1-7669 detects Rh(o)D and Rh(o)Dii human Rhesus system protein from human samples, with no cross-reactivity with other Rhesus antigens. MA1-7669 has been successfully used in agglutination applications.
Specifications
Specifications
| Antigen | RhoD |
| Applications | Agglutination |
| Classification | Monoclonal |
| Clone | 55/2/376 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1.25mM EDTA, 0.3M NaCl and 0.1% sodium azide; pH 7.2 |
| Gene | RHOD |
| Gene Accession No. | O00212 |
| Gene Alias | AI326383; aplysia ras-related homolog D; Arhd; LOW QUALITY PROTEIN: rho-related GTP-binding protein RhoD; ras homolog D; ras homolog D (RhoD); ras homolog family member D; ras homolog gene family, member A; ras homolog gene family, member D; Rh(o); Rh(o)ii; Rho; RHOD; RhoHP1; RHOM; Rho-related GTP-binding protein RhoD; Rho-related protein HP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
