Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Rhodamine 6G, 99%, pure

Catalog No. AC419010250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419010250 25 g Glass bottle
AC419010050 5 g Glass bottle
AC419011000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419010250 Supplier Thermo Scientific Chemicals Supplier No. 419010250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 989-38-8 | C28H31ClN2O3 | 479.017 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 989-38-8
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.017
MDL Number MFCD00012665
InChI Key VYXSBFYARXAAKO-UHFFFAOYSA-N
Synonym Basic Red 1, C.I. 45160
PubChem CID 51358423
IUPAC Name ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride
SMILES [H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-]

Specifications

Melting Point 290.0°C
Assay Percent Range 99%
Beilstein 18, V,12, 283
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 479.01
Percent Purity 99%
Grade Pure
Packaging Glass bottle
UV_Vis 98.5% min.
Color Brown to Brown/Red
Quantity 25 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Rhodamine 6G, 99%
Show More Show Less
missing translation for 'healthHazard2' GHS H Statement:
Harmful if swallowed.
missing translation for 'healthHazard3' GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-584-9
RTECSNumber DH0175000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.