Learn More
Abcam Rhodamine 6G Bis(oxyethylamino)ethane Amide Bis(trifluoroacetate), >95% Purity, 250 mg
Description
- Molecular weight: 772.73 g/mol
- Purity: >95%
- Application: Fluorescent labeling for proteomics research
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Label or Dye | Rhodamine 6G |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Product Type | Protein Labeling Reagent |
| Content And Storage | Store at ambient temperature. |
| Detection Method | Fluorescence |
| Formulation | 100% Rhodamine 6G bis(oxyethylamino)ethane amide bis(trifluoroacetate) |
| Quantity | 250 mg |
| For Use With (Application) | Proteomics research, Fluorescent labeling |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.