Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Rhodamine B Solution

Catalog No. 111016642 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6642 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6642 Supplier MilliporeSigma™ Supelco™ Supplier No. 02558500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

0.2% in isopropanol, for TLC derivatization

  • CAS: 81-88-9
  • Molecular Weight: 479.01
  • Density: 0.79 g/mL at 20°C
TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 mL
Type Rhodamine B
For Use With (Application) TLC Derivatization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.