Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium, 5% on alumina powder, C301099-5

Catalog No. AAH3620106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3620106 5 g
AAH3620114 25 g
AAH3620122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3620106 Supplier Thermo Scientific Chemicals Supplier No. H3620106
Add to Cart

CAS: 7440-16-6 | Rh | 102.91 g/mol

Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with organic materials and flammable substances.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]

Specifications

Color Gray
Physical Form Powder
Quantity 5 g
Solubility Information Insoluble in water.
Odor Odorless
Chemical Name or Material Rhodium, 5% on alumina powder, C301099-5
EINECSNumber 231-125-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.