Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rhodium(III) nitrate hydrate

Catalog No. 837501G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-750-1G 1 g
83-750-250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 83-750-1G Supplier Sigma Aldrich Supplier No. 837501G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Rhodium nitrate is commonly used as a precursor to synthesize rhodium.

Specifications

CAS 10139-58-9
Quantity 1 g
Molecular Formula N3O9Rh -+ xH2O
Linear Formula N3O9Rh -+ xH2O
MDL Number MFCD01862637
Synonym Rhodium trinitrate
Molecular Weight (g/mol) 288.92
Percent Purity ∼36%rhodium (Rh) basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.