Learn More
Cayman Chemical RhodojaponIn III 25mg
SDP
Supplier:  Cayman Chemical 3634525 MG
Encompass_Preferred
An inhibitor of H-PGDS (IC50 55 8 nM) selective for H-PGDS over L-PGDS at 100 UM as well as over enzyme and receptor panels at 10 UM inhibits A23187-induced PGD2 production in KU812 and RBL-2H3 basophils (IC50s 78 3 and 181 3 nM respectively) inhibits ovalbumin-induced nasal lavage fluid eosinophil infiltration and late-phase nasal obstruction in an ovalbumin-sensitized guinea pig model of allergic rhinitis at 30 mg/kg
Catalog No. 50-291-2201
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.