Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoGDI Polyclonal Antibody
GREENER_CHOICE

Catalog No. 511000Z
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-100-0Z 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-100-0Z Supplier Invitrogen™ Supplier No. 511000Z
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RhoGDI Polyclonal Antibody for ELISA, ICC/IF, IP, Western Blot

Aplysia Ras-related homologs (ARHs), also called Rho genes, belong to the RAS gene superfamily encoding small guanine nucleotide exchange (GTP/GDP) factors. The ARH proteins may be kept in the inactive, GDP-bound state by interaction with GDP dissociation inhibitors, such as ARHGDIA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoGDI
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene ARHGDIA
Gene Accession No. P19803, P52565, Q99PT1
Gene Alias 5330430M07Rik; Arhgdia; arhgdia {ECO:0000312; Bles01; C87222; epididymis secretory sperm binding protein Li 47e; Gdi1; Gdi-1; GDIA1; GDP-dissociation inhibitor, aplysia RAS-related 1; HEL-S-47e; MGC117248; NPHS8; QtrA-11619; RGD:1359547}; Rho GDI 1; rho GDI 1 {ECO:0000250; Rho GDI alpha; Rho GDP dissociation inhibitor (GDI) alpha; Rho GDP dissociation inhibitor alpha; rho GDP-dissociation inhibitor 1; rho GDP-dissociation inhibitor 1 {ECO:0000250; RhoDGI; RHOGDI; Rho-GDI; Rho-GDI alpha; rho-GDI alpha {ECO:0000250; RhoGDI-1; UniProtKB:P19803}; unnamed protein product
Gene Symbols ARHGDIA
Host Species Rabbit
Immunogen A 25 amino acid synthetic peptide derived from the N-terminus of the human Rho-GDI-alpha protein. This sequence is identical to the bovine sequence.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192662, 338054, 396
Target Species Human, Mouse, Bovine
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.