Learn More
RHOJ Mouse anti-Human, Clone: OTI1D7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505592

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ARHJ belongs to the Rho family of small GTP-binding proteins. Rho proteins regulate the dynamic assembly of cytoskeletal components for several physiologic processes, such as cell proliferation and motility and the establishment of cell polarity. They are also involved in pathophysiologic process, such as cell transformation and metastasis.Specifications
RHOJ | |
Monoclonal | |
Unconjugated | |
RHOJ | |
ARHJ, RASL7B, TC10B, TCL | |
Mouse | |
Affinity Chromatography | |
RUO | |
57381 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1D7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H4E5 | |
RHOJ | |
Full length human recombit protein of human RHOJ produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.